• USA Home
  • Search Results

  • 1butanol74127136311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Butanol

19 Products
Synonym:   1-Butanol, Butan-1-ol, n-Butyl alcohol, BuOH
Image of 1-Butanol
CAS Number:
Linear Formula:
 CH3(CH2)3OH
Molecular Weight:
 74.12
 EC Index Number:
 200-751-6
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Package quantity
  Restriction
  Refractive Index
  Anal. Standard Form
  UV-Absorption Condition
  Solubility
  Purity Grade
  pH Value
  Transition temperature
  Application(s)
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Transmittance
  Potency
  Physical Form
  Color
  UV-Absorption Values
  Vapor pressure
  Assay
Show More

Product #
Description
B7906 1-Butanol, for molecular biology, ≥99%,
281549 1-Butanol, anhydrous, 99.8%,
19422 1-Butanol, analytical standard,
360465 1-Butanol, ACS reagent, ≥99.4%,
34867 1-Butanol, for HPLC, ≥99.7%,
33065 1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC),
537993 1-Butanol, 99.9%,
154679 1-Butanol, spectrophotometric grade, 99.5%,
442665 1-Butanol, analytical standard,
1.01990 1-Butanol, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
270679 1-Butanol, for HPLC, 99.8%,
1.00988 1-Butanol, EMPROVE® ESSENTIAL NF,
1.01988 1-Butanol, for liquid chromatography LiChrosolv®,
8.22262 1-Butanol, EMPLURA®,
PHR1209 1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material,
1081807 1-Butanol, United States Pharmacopeia (USP) Reference Standard,
901351 1-Butanol, BioRenewable, ACS reagent, ≥99.4%, Green Alternative
900639 1-Butanol, anhydrous, ZerO2®, 99.8%,
RTC000092 1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material,