• USA Home
  • Search Results

  • 1butyl1methylpiperidiniumbistrifluoromethylsulfonylimide4364362358002911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Butyl-1-methylpiperidinium bis(trifluoromethylsulfonyl)imide

2 Products
Synonym:   PP14TFSI, [Bmpip][NTf2]
Image of 1-<WBR>Butyl-<WBR>1-<WBR>methylpiperidinium bis(trifluoromethylsulfonyl)<WBR>imide
CAS Number:
Empirical Formula (Hill Notation):
 C12H22F6N2O4S2
Molecular Weight:
 436.43
 Beilstein/REAXYS Number:
 10137193
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More