• USA Home
  • Search Results

  • 1butyl23dimethylimidazoliumtetrafluoroborate2400540284678011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Butyl-2,3-dimethylimidazolium tetrafluoroborate

2 Products
Image of 1-<WBR>Butyl-<WBR>2,3-<WBR>dimethylimidazolium tetrafluoroborate
CAS Number:
Empirical Formula (Hill Notation):
 C9H17BF4N2
Molecular Weight:
 240.05
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More