• USA Home
  • Search Results

  • 1chloro24dinitrobenzene202559700711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Chloro-2,4-dinitrobenzene

3 Products
Synonym:   2,4-Dinitrochlorobenzene, CDNB, DNCB
Image of 1-<WBR>Chloro-<WBR>2,4-<WBR>dinitrobenzene
CAS Number:
Linear Formula:
 ClC6H3(NO2)2
Molecular Weight:
 202.55
 Beilstein/REAXYS Number:
 613161
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Potency
  Assay
Show More