• USA Home
  • Search Results

  • 1chloro4iodobenzene2384563787611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Chloro-4-iodobenzene

1 Products
Image of 1-<WBR>Chloro-<WBR>4-<WBR>iodobenzene
CAS Number:
Linear Formula:
 ClC6H4I
Molecular Weight:
 238.45
 Beilstein Registry Number:
 1634414
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More