• USA Home
  • Search Results

  • 1chloronaphthalene162629013111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Chloronaphthalene

2 Products
Image of 1-Chloronaphthalene
CAS Number:
Empirical Formula (Hill Notation):
 C10H7Cl
Molecular Weight:
 162.62
 Beilstein/REAXYS Number:
 970836
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Refractive Index
  Potency
  Density
  Assay
Show More