• USA Home
  • Search Results

  • 1decanol1582811230111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Decanol

6 Products
Synonym:   n-Decyl alcohol, Alcohol C10
Image of 1-Decanol
CAS Number:
Linear Formula:
 CH3(CH2)9OH
Molecular Weight:
 158.28
 Beilstein/REAXYS Number:
 1735221
 Council of Europe no.:
 73
 EC Number:
 FEMA Number:
 2365
 Flavis number:
 2.024
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Titertest
  Potency
  Special Grade
  Assay
Show More