• USA Home
  • Search Results

  • 1deoxy1morpholinodfructose24926629116311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Deoxy-1-morpholino-D-fructose

1 Products
Image of 1-<WBR>Deoxy-<WBR>1-<WBR>morpholino-<SC>D</SC>-fructose
CAS Number:
Empirical Formula (Hill Notation):
 C10H19NO6
Molecular Weight:
 249.26
 Beilstein/REAXYS Number:
 19582
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More