• USA Home
  • Search Results

  • 1dodecanethiol2024011255011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Dodecanethiol

3 Products
Synonym:   n-Dodecyl mercaptan, Dodecyl mercaptan, Lauryl mercaptan, Mercaptan C12, NDM
Image of 1-Dodecanethiol
CAS Number:
Linear Formula:
 CH3(CH2)11SH
Molecular Weight:
 202.40
 Beilstein/REAXYS Number:
 969337
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More