• USA Home
  • Search Results

  • 1ethyl3methylimidazoliumbistrifluoromethylsulfonylimide3913117489982211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

3 Products
Synonym:   EMIM BTI, EMIM TFSI, EMIMIm
Image of 1-<WBR>Ethyl-<WBR>3-<WBR>methylimidazolium bis(trifluoromethylsulfonyl)<WBR>imide
CAS Number:
Empirical Formula (Hill Notation):
 C8H11F6N3O4S2
Molecular Weight:
 391.31
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More