• USA Home
  • Search Results

  • 1ethyl3methylimidazoliumchloride146626503909011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Ethyl-3-methylimidazolium chloride

5 Products
Synonym:   EMIC
Image of 1-<WBR>Ethyl-<WBR>3-<WBR>methylimidazolium chloride
CAS Number:
Empirical Formula (Hill Notation):
 C6H11ClN2
Molecular Weight:
 146.62
 Beilstein/REAXYS Number:
 5163190
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More