• USA Home
  • Search Results

  • 1ethyl3methylimidazoliumtetrachloroaluminate279968043205911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Ethyl-3-methylimidazolium tetrachloroaluminate

2 Products
Synonym:   1-Ethyl-3-methyl-1H-imidazolium tetrachloroaluminate
Image of 1-<WBR>Ethyl-<WBR>3-<WBR>methylimidazolium tetrachloroaluminate
CAS Number:
Empirical Formula (Hill Notation):
 C6H11AlCl4N2
Molecular Weight:
 279.96
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More