• USA Home
  • Search Results

  • 1ethyl3methylimidazoliumthiocyanate1692533171763611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Ethyl-3-methylimidazolium thiocyanate

3 Products
Synonym:   1-Ethyl-3-methylimidazolium thiocyanate
Image of 1-<WBR>Ethyl-<WBR>3-<WBR>methylimidazolium thiocyanate
CAS Number:
Empirical Formula (Hill Notation):
 C7H11N3S
Molecular Weight:
 169.25
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More