• USA Home
  • Search Results

  • 1ethyl4ethynylbenzene130194030711711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Ethyl-4-ethynylbenzene

1 Products
Image of 1-<WBR>Ethyl-<WBR>4-<WBR>ethynylbenzene
CAS Number:
Linear Formula:
 CH3CH2C6H4C≡CH
Molecular Weight:
 130.19
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More