• USA Home
  • Search Results

  • 1ethylpiperazine11419530825811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Ethylpiperazine

1 Products
Image of 1-Ethylpiperazine
CAS Number:
Empirical Formula (Hill Notation):
 C6H14N2
Molecular Weight:
 114.19
 Beilstein/REAXYS Number:
 102971
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
415308 1-Ethylpiperazine, 98%,