• USA Home
  • Search Results

  • 1ethynylcyclohexene1061793149711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Ethynylcyclohexene

1 Products
Image of 1-Ethynylcyclohexene
CAS Number:
Linear Formula:
 C6H9C≡CH
Molecular Weight:
 106.17
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
316571 1-Ethynylcyclohexene, 99%,