• USA Home
  • Search Results

  • 1ethynylcyclohexylamine123203038918511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Ethynylcyclohexylamine

1 Products
Image of 1-<WBR>Ethynylcyclohexylamine
CAS Number:
Linear Formula:
 HC≡CC6H10NH2
Molecular Weight:
 123.20
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More