• USA Home
  • Search Results

  • 1ethynylnaphthalene152191572765811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Ethynylnaphthalene

1 Products
Image of 1-Ethynylnaphthalene
CAS Number:
Linear Formula:
 C10H7C≡CH
Molecular Weight:
 152.19
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
557927 1-Ethynylnaphthalene, 97%,