• USA Home
  • Search Results

  • 1formylpiperidine11316259186811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Formylpiperidine

1 Products
Synonym:   N-Carbaldehyde piperidine, NFP, Piperidine-1-carboxaldehyde
Image of 1-Formylpiperidine
CAS Number:
Empirical Formula (Hill Notation):
 C6H11NO
Molecular Weight:
 113.16
 Beilstein/REAXYS Number:
 107697
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
F17407 1-Formylpiperidine, 99%,