• USA Home
  • Search Results

  • 1h1h2h2hperfluorooctyliodide47400204357411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1H,1H,2H,2H-Perfluorooctyl iodide

1 Products
Synonym:   1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 1H,1H,2H,2H-Perfluorooctyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl iodide, 8-Iodo-1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorooctane
Image of 1<I>H</I>,1<I>H</I>,2<I>H</I>,2<I>H</I>-Perfluorooctyl iodide
CAS Number:
Linear Formula:
 CF3(CF2)5CH2CH2I
Molecular Weight:
 474.00
 Beilstein/REAXYS Number:
 1887105
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More