• USA Home
  • Search Results

  • 1heptanethiol13227163909411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Heptanethiol

3 Products
Image of 1-Heptanethiol
CAS Number:
Empirical Formula (Hill Notation):
 C7H16S
Molecular Weight:
 132.27
 EC Index Number
 216-678-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More