• USA Home
  • Search Results

  • 1hexacosanol3827150652511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Hexacosanol

1 Products
Synonym:   Ceryl alcohol, Hexacosyl alcohol
Image of 1-Hexacosanol
CAS Number:
Linear Formula:
 CH3(CH2)25OH
Molecular Weight:
 382.71
 Beilstein/REAXYS Number:
 1783162
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More