• USA Home
  • Search Results

  • 1hexadecanol242443665382411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Hexadecanol

3 Products
Synonym:   Cetyl alcohol, Palmityl alcohol
Image of 1-Hexadecanol
CAS Number:
Linear Formula:
 CH3(CH2)15OH
Molecular Weight:
 242.44
 Beilstein/REAXYS Number:
 1748475
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Physical Form
Show More