• USA Home
  • Search Results

  • 1hexadecene2244362973211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Hexadecene

3 Products
Synonym:   Cetene
Image of 1-Hexadecene
CAS Number:
Linear Formula:
 CH3(CH2)13CH=CH2
Molecular Weight:
 224.43
 Beilstein/REAXYS Number:
 1753401
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Physical Form
  Potency
  Special Grade
  Application(s)
Show More