• USA Home
  • Search Results

  • 1hexen3ol10016479844111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Hexen-3-ol

2 Products
Image of 1-Hexen-3-ol
CAS Number:
Linear Formula:
 CH3CH2CH2CH(OH)CH=CH2
Molecular Weight:
 100.16
 Council of Europe no.:
 10220
 EC Number:
 FEMA Number:
 3608
 Flavis number:
 2.104
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More