• USA Home
  • Search Results

  • 1hexyl3methylimidazoliumchloride2027217105817611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Hexyl-3-methylimidazolium chloride

2 Products
Synonym:   HMIMCl
Image of 1-<WBR>Hexyl-<WBR>3-<WBR>methylimidazolium chloride
CAS Number:
Empirical Formula (Hill Notation):
 C10H19ClN2
Molecular Weight:
 202.72
 Beilstein/REAXYS Number:
 8356525
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More