• USA Home
  • Search Results

  • 1hexyne821469302711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Hexyne

2 Products
Image of 1-Hexyne
CAS Number:
Linear Formula:
 CH3(CH2)3C≡CH
Molecular Weight:
 82.14
 Beilstein/REAXYS Number:
 635687
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39010407
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More