• USA Home
  • Search Results

  • 1hydroxypyrene21825531579711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Hydroxypyrene

2 Products
Synonym:   1-Pyrenol
Image of 1-Hydroxypyrene
CAS Number:
Empirical Formula (Hill Notation):
 C16H10O
Molecular Weight:
 218.25
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More