• USA Home
  • Search Results

  • 1iodo22dimethylpropane198051550133411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Iodo-2,2-dimethylpropane

1 Products
Synonym:   2,2-Dimethyl-1-iodopropane, Neopentyl iodide
Image of 1-<WBR>Iodo-<WBR>2,2-<WBR>dimethylpropane
CAS Number:
Linear Formula:
 (CH3)3CCH2I
Molecular Weight:
 198.05
 Beilstein/REAXYS Number:
 1718796
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More