• USA Home
  • Search Results

  • 1methyl3methoxy4isopropylbenzene16424107656811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Methyl-3-methoxy-4-isopropylbenzene

1 Products
Synonym:   2-Isopropyl-5-methylanisole, O-Methylthymol, NSC 404221, Thymol methyl ether
Image of 1-<WBR>Methyl-<WBR>3-<WBR>methoxy-<WBR>4-<WBR>isopropylbenzene
CAS Number:
Empirical Formula (Hill Notation):
 C11H16O
Molecular Weight:
 164.24
 Council of Europe no.:
 11245
 EC Number:
 FEMA Number:
 3436
 Flavis number:
 4.043
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More