• USA Home
  • Search Results

  • 1methyl3trifluoromethyl1hpyrazole4boronicacidpinacolester27606121879053411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Methyl-3-trifluoromethyl-1H-pyrazole-4-boronic acid pinacol ester

2 Products
Synonym:   1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)-1H-pyrazole
Image of 1-<WBR>Methyl-<WBR>3-<WBR>trifluoromethyl-<WBR>1<I>H</I>-pyrazole-4-boronic acid pinacol ester
CAS Number:
Empirical Formula (Hill Notation):
 C11H16BF3N2O2
Molecular Weight:
 276.06
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More