• USA Home
  • Search Results

  • 1methyl4phenyl1236tetrahydropyridinehydrochloride209722300785411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride

1 Products
Synonym:   MPTP hydrochloride
Image of 1-<WBR>Methyl-<WBR>4-<WBR>phenyl-<WBR>1,2,3,6-<WBR>tetrahydropyridine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C12H15N · HCl
Molecular Weight:
 209.72
 Beilstein/REAXYS Number:
 3707312
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More