• USA Home
  • Search Results

  • 1methyluricacid1821470879211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Methyluric acid

1 Products
Synonym:   1-Methyl-2,6,8-trihydroxypurine, 1-U, 1MUA
Image of 1-Methyluric acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H6N4O3
Molecular Weight:
 182.14
 Beilstein/REAXYS Number:
 195190
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
M6885 1-Methyluric acid,