• USA Home
  • Search Results

  • 1naphthol144179015311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Naphthol

8 Products
Synonym:   α-Naphthol, 1-Hydroxynaphthalene
Image of 1-Naphthol
CAS Number:
Linear Formula:
 C10H7OH
Molecular Weight:
 144.17
 Beilstein/REAXYS Number:
 1817321
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Titertest
  Product Line
  Vapor pressure
  Transition temperature
  Solubility
  Purity Grade
  Density
  Bulk Density
  Autoignition temp
  Potency
  Melting Point
  Special Grade
Show More