• USA Home
  • Search Results

  • 1naphthoylchloride1906387918511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Naphthoyl chloride

1 Products
Image of 1-Naphthoyl chloride
CAS Number:
Linear Formula:
 C10H7COCl
Molecular Weight:
 190.63
 Beilstein/REAXYS Number:
 775785
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
250252 1-Naphthoyl chloride, 97%,