• USA Home
  • Search Results

  • 1naphthylamine1431913432711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Naphthylamine

3 Products
Synonym:   α-Naphthylamine, 1-Aminonaphthalene
Image of 1-Naphthylamine
CAS Number:
Linear Formula:
 C10H7NH2
Molecular Weight:
 143.19
 Beilstein/REAXYS Number:
 386133
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
Show More