• USA Home
  • Search Results

  • 1naphthylmethylamine1572111831011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Naphthylmethylamine

1 Products
Synonym:   1-(Aminomethyl)naphthalene, 1-Naphthalenemethylamine
Image of 1-<WBR>Naphthylmethylamine
CAS Number:
Linear Formula:
 C10H7CH2NH2
Molecular Weight:
 157.21
 Beilstein/REAXYS Number:
 2206459
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More