• USA Home
  • Search Results

  • 1nonene1262412411811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Nonene

2 Products
Image of 1-Nonene
CAS Number:
Linear Formula:
 CH3(CH2)6CH=CH2
Molecular Weight:
 126.24
 Beilstein/REAXYS Number:
 1698439
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More