• USA Home
  • Search Results

  • 1octadecanol2704911292511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Octadecanol

3 Products
Synonym:   Octadecyl alcohol, Stearyl alcohol
Image of 1-Octadecanol
CAS Number:
Linear Formula:
 CH3(CH2)17OH
Molecular Weight:
 270.49
 Beilstein/REAXYS Number:
 1362907
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Solubility
Show More