• USA Home
  • Search Results

  • 1octanol1302311187511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Octanol

9 Products
Synonym:   Alcohol C8, Capryl alcohol, Octyl alcohol
Image of 1-Octanol
CAS Number:
Linear Formula:
 CH3(CH2)7OH
Molecular Weight:
 130.23
 Beilstein/REAXYS Number:
 1697461
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Refractive Index
  Document Links
  Allergen
  Organoleptic property
  Viscosity
  Vapor pressure
  UV-Absorption Condition
  Transition temperature
  Product Line
  Density
  Autoignition temp
  Solubility
  Potency
  Color
  Application(s)
  Agency/Method
  UV-Absorption Values
  Explosion limit
  Physical Form
  Special Grade
Show More