• USA Home
  • Search Results

  • 1octen3ol12821339186411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Octen-3-ol

4 Products
Synonym:   Pentyl vinyl carbinol
Image of 1-Octen-3-ol
CAS Number:
Linear Formula:
 CH3(CH2)4CH(OH)CH=CH2
Molecular Weight:
 128.21
 Beilstein/REAXYS Number:
 1744110
 Council of Europe no.:
 72
 EC Number:
 FEMA Number:
 2805
 Flavis number:
 2.023
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Application(s)
  Potency
  Agency/Method
  Special Grade
  Assay
Show More