• USA Home
  • Search Results

  • 1octyn3ol1262081872411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Octyn-3-ol

1 Products
Image of 1-Octyn-3-ol
CAS Number:
Linear Formula:
 CH3(CH2)4CH(OH)C≡CH
Molecular Weight:
 126.20
 Beilstein/REAXYS Number:
 1098642
 EC Number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
127280 1-Octyn-3-ol, 96%,