• USA Home
  • Search Results

  • 1octyne1102062905011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Octyne

1 Products
Image of 1-Octyne
CAS Number:
Linear Formula:
 CH3(CH2)5C≡CH
Molecular Weight:
 110.20
 Beilstein/REAXYS Number:
 1734494
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39010411
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
244465 1-Octyne, 97%,