• USA Home
  • Search Results

  • 1oleoylracglycerol3565411103511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Oleoyl-rac-glycerol

2 Products
Synonym:   1-(cis-9-Octadecenoyl)-rac-glycerol, rac-Glycerol 1-monooleate, DL-α-Monoolein, Glyceryl cis-9-octadecenoate, MG(18:1(9Z)/0:0/0:0)[rac], Monoolein
Image of 1-Oleoyl-<I>rac</I>-glycerol
CAS Number:
Linear Formula:
 CH3(CH2)7CH=CH(CH2)7COOCH2CHOHCH2OH
Molecular Weight:
 356.54
 Beilstein/REAXYS Number:
 1728976
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  ID clarifier
Show More