• USA Home
  • Search Results

  • 1pentadecene210401336061711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Pentadecene

1 Products
Image of 1-Pentadecene
CAS Number:
Linear Formula:
 CH3(CH2)12CH=CH2
Molecular Weight:
 210.40
 Beilstein/REAXYS Number:
 1751011
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
222887 1-Pentadecene, 98%,