• USA Home
  • Search Results

  • 1pentanol88157141011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Pentanol

6 Products
Synonym:   n-Amyl alcohol, Pentyl alcohol
Image of 1-Pentanol
CAS Number:
Linear Formula:
 CH3(CH2)4OH
Molecular Weight:
 88.15
 Beilstein/REAXYS Number:
 1730975
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Refractive Index
  Anal. Standard Form
  Autoignition temp
  pH Value
  Application(s)
  Assay
  Physical Form
Show More