• USA Home
  • Search Results

  • 1penten3one8412162958911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Penten-3-one

2 Products
Synonym:   Ethyl vinyl ketone
Image of 1-Penten-3-one
CAS Number:
Linear Formula:
 C2H5COCH=CH2
Molecular Weight:
 84.12
 Beilstein/REAXYS Number:
 1735857
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Additive
  Titertest
Show More