• USA Home
  • Search Results

  • 1pentene701310967111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Pentene

3 Products
Image of 1-Pentene
CAS Number:
Linear Formula:
 CH3CH2CH2CH=CH2
Molecular Weight:
 70.13
 Beilstein/REAXYS Number:
 1731629
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Refractive Index
  Application(s)
  Assay
Show More