• USA Home
  • Search Results

  • 1phenyl2propyn1ol13216418787511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Phenyl-2-propyn-1-ol

1 Products
Synonym:   (±)-α-Ethynylbenzyl alcohol, (±)-1-Phenyl-2-propyn-1-ol, (±)-3-Hydroxy-3-phenyl-1-propyne, 1-Phenylpropargyl alcohol
Image of 1-<WBR>Phenyl-<WBR>2-<WBR>propyn-<WBR>1-<WBR>ol
CAS Number:
Linear Formula:
 CH≡CCH(OH)C6H5
Molecular Weight:
 132.16
 Beilstein/REAXYS Number:
 742365
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More