• USA Home
  • Search Results

  • 1phenyl2trimethylsilylacetylene17431217006111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Phenyl-2-trimethylsilylacetylene

1 Products
Synonym:   1-(Trimethylsilyl)-2-phenylacetylene, Phenylethynyl-trimethylsilane, Trimethyl(phenylethynyl)silane
Image of 1-<WBR>Phenyl-<WBR>2-<WBR>trimethylsilylacetylene
CAS Number:
Linear Formula:
 C6H5C≡CSi(CH3)3
Molecular Weight:
 174.31
 Beilstein/REAXYS Number:
 3088678
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More